Assay Details
Target Gene Details
Entrez Gene ID: | 104326052 |
Gene Name: | negative regulator of interferon response (non-protein coding) |
Gene Aliases: |
lncRNA-CMPK2 |
Location: |
Chr.2:6828554-6840464 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRIR | NR_126359.1 | |||
BP421873.1 | ||||
BQ437827.1 |
