Assay Details
Target Gene Details
Entrez Gene ID: | 79901 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
CYB561A2, DCYTB, FRRS3 |
Location: |
Chr.2:171522247-171558133 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYBRD1 | NM_001127383.1 | NP_001120855.1 | ||
NM_001256909.1 | NP_001243838.1 | |||
NM_024843.3 | NP_079119.3 | |||
AK027115.1 | BAB15661.1 | |||
AK128495.1 | BAC87466.1 | |||
AK225459.1 | ||||
AK291731.1 | ||||
AK301485.1 | ||||
AK316588.1 | ||||
AL136693.1 | CAB66628.1 | |||
BC065290.1 | AAH65290.1 | |||
CX866825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160884 | Chr.2:171536097 - 171553747 on Build GRCh38 | Duplication |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
