Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANSL1L | NM_001307976.1 | NP_001294905.1 | ||
| NM_152519.3 | NP_689732.2 | |||
| XM_005246328.4 | XP_005246385.1 | |||
| XM_005246329.4 | XP_005246386.1 | |||
| XM_005246330.4 | XP_005246387.1 | |||
| XM_005246332.4 | XP_005246389.1 | |||
| XM_011510706.2 | XP_011509008.1 | |||
| XM_011510707.2 | XP_011509009.1 | |||
| XM_011510710.2 | XP_011509012.1 | |||
| XM_017003430.1 | XP_016858919.1 | |||
| XM_017003431.1 | XP_016858920.1 | |||
| AK056911.1 | BAB71308.1 | |||
| AK074441.1 | BAB85085.1 | |||
| AL133053.1 | CAB61379.1 | |||
| BC032837.1 | AAH32837.1 | |||
| BC126155.1 | ||||
| BC143914.1 |
Target Gene Details
Entrez Gene ID: | 101928020 |
Gene Name: | uncharacterized LOC101928020 |
Gene Aliases: |
- |
Location: |
Chr.2:210029922-210064356 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928020 | NR_110291.1 | |||
| AK093026.1 |

Genomic Map