Assay Details
Target Gene Details
Entrez Gene ID: | 9270 |
Gene Name: | integrin subunit beta 1 binding protein 1 |
Gene Aliases: |
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B |
Location: |
Chr.2:9405681-9423569 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
| nsv2602 | Chr.2:9408817 - 9453535 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map