Assay Details
Target Gene Details
Entrez Gene ID: | 51626 |
Gene Name: | dynein cytoplasmic 2 light intermediate chain 1 |
Gene Aliases: |
CGI-60, D2LIC, LIC3 |
Location: |
Chr.2:43774039-43828492 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNC2LI1 | NM_001193464.1 | NP_001180393.1 | ||
| NM_015522.3 | NP_056337.1 | |||
| NM_016008.3 | NP_057092.2 | |||
| XM_005264364.4 | XP_005264421.1 | |||
| XM_005264365.4 | XP_005264422.1 | |||
| AF151818.1 | AAD34055.1 | |||
| AK074206.1 | ||||
| AK223432.1 | BAD97152.1 | |||
| AK301489.1 | ||||
| AL050006.1 | CAB43233.1 | |||
| AY083346.1 | AAL99216.1 | |||
| BC006969.1 | AAH06969.1 | |||
| BC016324.1 | AAH16324.1 | |||
| BC040558.1 | AAH40558.1 | |||
| BC058823.1 | AAH58823.1 | |||
| BG716986.1 | ||||
| BX493698.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891993 | Chr.2:43470750 - 43815292 on Build GRCh38 | Gain |
|
| nsv1012126 | Chr.2:43716882 - 44206414 on Build GRCh38 | Gain |
|
| nsv998912 | Chr.2:43780254 - 43835757 on Build GRCh38 | Gain |
|
| dgv59n68 | Chr.2:43619845 - 43871585 on Build GRCh38 | Gain |
|
| nsv1007740 | Chr.2:42961811 - 44117003 on Build GRCh38 | Gain |
|
| nsv581661 | Chr.2:43762402 - 44145770 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150856355] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map