Assay Details
Target Gene Details
Entrez Gene ID: | 5136 |
Gene Name: | phosphodiesterase 1A |
Gene Aliases: |
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A |
Location: |
Chr.2:182140035-182523375 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE1A | NM_001258312.1 | 16 | 4847 | NP_001245241.1 |
| NM_005019.4 | 15 | 4862 | NP_005010.2 | |
| XM_017004302.1 | 15 | 4994 | XP_016859791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593559 | Chr.2:182126607 - 182163512 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map