Assay Details
Target Gene Details
Entrez Gene ID: | 285148 |
Gene Name: | isoamyl acetate-hydrolyzing esterase 1 homolog |
Gene Aliases: |
- |
Location: |
Chr.2:9473638-9512403 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IAH1 | NM_001039613.2 | NP_001034702.1 | ||
| NM_001320858.1 | NP_001307787.1 | |||
| NM_001320859.1 | NP_001307788.1 | |||
| NM_001320860.1 | NP_001307789.1 | |||
| NM_001320863.1 | NP_001307792.1 | |||
| XM_017003878.1 | XP_016859367.1 | |||
| AK092982.1 | ||||
| AK291984.1 | ||||
| AK297648.1 | ||||
| BC019061.1 | ||||
| BC059410.1 | AAH59410.1 | |||
| BC067865.1 | AAH67865.1 | |||
| BC111025.1 | AAI11026.1 | |||
| BF447213.1 | ||||
| BM553525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map