Assay Details
Target Gene Details
Entrez Gene ID: | 659 |
Gene Name: | bone morphogenetic protein receptor type 2 |
Gene Aliases: |
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK |
Location: |
Chr.2:202376310-202567751 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMPR2 | NM_001204.6 | 1 | 322 | NP_001195.2 |
| XM_011511687.1 | 1 | 339 | XP_011509989.1 | |
| BU176845.1 | 1 | 279 | ||
| DA787547.1 | 1 | 322 | ||
| DB233604.1 | 1 | 155 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007686 | Chr.2:202370673 - 202421699 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map