Assay Details
Target Gene Details
Entrez Gene ID: | 116987 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 1 |
Gene Aliases: |
AGAP-1, CENTG2, GGAP1, cnt-g2 |
Location: |
Chr.2:235494089-236131800 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP1 | NM_001037131.2 | NP_001032208.1 | ||
| NM_001244888.1 | NP_001231817.1 | |||
| NM_014914.4 | NP_055729.2 | |||
| XM_005246059.4 | XP_005246116.1 | |||
| XM_006712234.3 | XP_006712297.1 | |||
| XM_006712235.3 | XP_006712298.1 | |||
| AB029022.1 | ||||
| AF413078.1 | AAL04172.1 | |||
| AY033765.1 | AAK56506.1 | |||
| BC060814.1 | AAH60814.1 | |||
| BC140856.1 | ||||
| BC167153.1 | ||||
| BC167157.1 | ||||
| KJ534764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527654 | Chr.2:234661964 - 235549244 on Build GRCh38 | Gain |
|
| nsv834575 | Chr.2:235384329 - 235543362 on Build GRCh38 | Gain |
|
| esv3575341 | Chr.2:235300440 - 235588493 on Build GRCh38 | Gain |
|
| dgv4191n100 | Chr.2:235085672 - 235523486 on Build GRCh38 | Gain |
|
| nsv999839 | Chr.2:234676935 - 235726730 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map