Assay Details
Target Gene Details
Entrez Gene ID: | 8809 |
Gene Name: | interleukin 18 receptor 1 |
Gene Aliases: |
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP |
Location: |
Chr.2:102355817-102398777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18R1 | NM_001282399.1 | NP_001269328.1 | ||
| NM_003855.3 | NP_003846.1 | |||
| XM_017005182.1 | XP_016860671.1 | |||
| XM_017005183.1 | XP_016860672.1 | |||
| XM_017005184.1 | XP_016860673.1 | |||
| DR006178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160830 | Chr.2:102344124 - 102357571 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map