Assay Details
Target Gene Details
Entrez Gene ID: | 25801 |
Gene Name: | grancalcin |
Gene Aliases: |
GCL |
Location: |
Chr.2:162318768-162371605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCA | XM_006712398.3 | XP_006712461.1 | ||
| XM_006712400.3 | XP_006712463.1 | |||
| XM_006712401.3 | XP_006712464.1 | |||
| XM_017003764.1 | XP_016859253.1 | |||
| XM_017003765.1 | XP_016859254.1 | |||
| XM_017003766.1 | XP_016859255.1 | |||
| XM_017003769.1 | XP_016859258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map