Assay Details
Target Gene Details
Entrez Gene ID: | 6160 |
Gene Name: | ribosomal protein L31 |
Gene Aliases: |
L31 |
Location: |
Chr.2:101002229-101019693 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL31 | NM_000993.4 | NP_000984.1 | ||
| NM_001098577.2 | NP_001092047.1 | |||
| NM_001099693.1 | NP_001093163.1 | |||
| AK297167.1 | ||||
| AK297247.1 | ||||
| AK297483.1 | ||||
| AK311775.1 | ||||
| BC017343.2 | AAH17343.1 | |||
| BC070210.1 | AAH70210.1 | |||
| BE564858.1 | ||||
| BG031488.1 | ||||
| CD247800.1 | ||||
| CF124259.1 | ||||
| CN269912.1 | ||||
| DA869102.1 | ||||
| X15940.1 | CAA34066.1 | |||
| X69181.1 | CAA48925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv961844 | Chr.2:101002623 - 101006193 on Build GRCh38 | Duplication |
|
| nsv834316 | Chr.2:100916586 - 101115039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map