Assay Details
Target Gene Details
Entrez Gene ID: | 101669764 |
Gene Name: | GPR1 antisense RNA |
Gene Aliases: |
GPR1-AS1, GPR1AS |
Location: |
Chr.2:206203376-206266243 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR1-AS | NR_104359.1 | |||
| AB774455.1 | ||||
| BE734382.1 |

Genomic Map