Assay Details
Target Gene Details
Entrez Gene ID: | 6326 |
Gene Name: | sodium voltage-gated channel alpha subunit 2 |
Gene Aliases: |
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2 |
Location: |
Chr.2:165208056-165392310 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN2A | XM_017004656.1 | XP_016860145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2082n106 | Chr.2:165221625 - 165222143 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map