Assay Details
Target Gene Details
Entrez Gene ID: | 100505876 |
Gene Name: | CEBPZ opposite strand |
Gene Aliases: |
CEBPZ-AS1 |
Location: |
Chr.2:37196451-37216193 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZOS | NM_001322373.1 | NP_001309302.1 | ||
| NM_001322374.1 | NP_001309303.1 | |||
| NM_001322375.1 | NP_001309304.1 | |||
| NM_001322376.1 | NP_001309305.1 | |||
| NR_136316.1 | ||||
| XM_017003106.1 | XP_016858595.1 | |||
| XM_017003107.1 | XP_016858596.1 | |||
| XM_017003108.1 | XP_016858597.1 | |||
| XM_017003109.1 | XP_016858598.1 | |||
| AK001814.1 | ||||
| BM838731.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79475241] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map