Assay Details
Target Gene Details
Entrez Gene ID: | 375298 |
Gene Name: | ceramide kinase like |
Gene Aliases: |
RP26 |
Location: |
Chr.2:181536672-181657107 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERKL | NM_001030311.2 | NP_001025482.1 | ||
| NM_001030312.2 | NP_001025483.1 | |||
| NM_001030313.2 | NP_001025484.1 | |||
| NM_001160277.1 | NP_001153749.1 | |||
| NM_201548.4 | NP_963842.1 | |||
| NR_027689.1 | ||||
| NR_027690.1 | ||||
| AJ640141.1 | CAG26695.1 | |||
| AJ697855.1 | CAG26977.1 | |||
| AJ697856.1 | CAG26978.1 | |||
| AJ697858.1 | CAG26980.1 | |||
| AK129976.1 | BAC85266.1 | |||
| AK293844.1 | ||||
| AY357073.1 | AAR13670.1 | |||
| AY690329.1 | AAW47988.1 | |||
| AY690330.1 | AAW47989.1 | |||
| AY690331.1 | AAW47990.1 | |||
| AY690332.1 | AAW47991.1 | |||
| AY690333.1 | AAW47992.1 | |||
| BC137498.1 | ||||
| BC137499.1 | ||||
| HQ426666.1 | ||||
| HQ426667.1 | ||||
| HQ426668.1 | ||||
| HQ426669.1 | ||||
| HQ426670.1 | ||||
| HQ426671.1 | ||||
| HQ426672.1 | ||||
| HQ426673.1 | ||||
| HQ426674.1 | ||||
| HQ426675.1 | ||||
| HQ426676.1 | ||||
| HQ426677.1 | ||||
| HQ426678.1 | ||||
| HQ426679.1 | ||||
| JF290420.1 |
More Information
Set Membership: |
|

Genomic Map