Assay Details
Target Gene Details
Entrez Gene ID: | 5624 |
Gene Name: | protein C, inactivator of coagulation factors Va and VIIIa |
Gene Aliases: |
APC, PC, PROC1, THPH3, THPH4 |
Location: |
Chr.2:127418221-127429246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROC | NM_000312.3 | NP_000303.1 | ||
| XM_005263717.4 | XP_005263774.1 | |||
| XM_017004505.1 | XP_016859994.1 | |||
| AK298280.1 | ||||
| AK298449.1 | ||||
| AK298454.1 | ||||
| AK303773.1 | ||||
| BC034377.1 | AAH34377.1 | |||
| CB113383.1 | ||||
| DA642156.1 | ||||
| X02750.1 | CAA26528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583036 | Chr.2:127416243 - 127426338 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map