Assay Details
Target Gene Details
Entrez Gene ID: | 84168 |
Gene Name: | anthrax toxin receptor 1 |
Gene Aliases: |
ATR, GAPO, TEM8 |
Location: |
Chr.2:69013144-69249327 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANTXR1 | NM_018153.3 | NP_060623.2 | ||
NM_032208.2 | NP_115584.1 | |||
NM_053034.2 | NP_444262.1 | |||
XM_017005075.1 | XP_016860564.1 | |||
XM_017005076.1 | XP_016860565.1 | |||
XM_017005077.1 | XP_016860566.1 | |||
AF279145.2 | AAK52094.1 | |||
AF421380.1 | AAL26496.1 | |||
AK001463.1 | BAA91707.1 | |||
AK223273.1 | BAD96993.1 | |||
AK292113.1 | ||||
AK309553.1 | ||||
BC012074.1 | AAH12074.1 | |||
CF126791.1 | ||||
DC360415.1 | ||||
DC361072.1 | ||||
JX424838.1 | ||||
JX424839.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
