Assay Details
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003130 | Chr.2:171332686 - 171380758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map