Assay Details
Target Gene Details
Entrez Gene ID: | 101929130 |
Gene Name: | uncharacterized LOC101929130 |
Gene Aliases: |
- |
Location: |
Chr.3:187291272-187297933 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929130 | NR_135551.1 | |||
BX648496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009271 | Chr.3:187246858 - 187480081 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
