Assay Details
Target Gene Details
Entrez Gene ID: | 1232 |
Gene Name: | C-C motif chemokine receptor 3 |
Gene Aliases: |
CC-CKR-3, CD193, CKR3, CMKBR3 |
Location: |
Chr.3:46210699-46266706 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR3 | XM_006712960.3 | XP_006713023.1 | ||
| XM_011533335.2 | XP_011531637.1 | |||
| XM_017005686.1 | XP_016861175.1 |
More Information
Set Membership: |
|

Genomic Map