Assay Details
Target Gene Details
Entrez Gene ID: | 55207 |
Gene Name: | ADP ribosylation factor like GTPase 8B |
Gene Aliases: |
ARL10C, Gie1 |
Location: |
Chr.3:5122245-5180916 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL8B | NM_018184.2 | NP_060654.1 | ||
| AB118751.1 | BAD23992.1 | |||
| AK001564.1 | BAA91759.1 | |||
| AK295465.1 | ||||
| AK298951.1 | ||||
| AK303872.1 | ||||
| BC013131.1 | AAH13131.1 | |||
| BC063125.1 | AAH63125.1 | |||
| CR457264.1 | CAG33545.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77454822] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map