Assay Details
Target Gene Details
Entrez Gene ID: | 151903 |
Gene Name: | coiled-coil domain containing 12 |
Gene Aliases: |
- |
Location: |
Chr.3:46917905-46982031 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC12 | NM_001277074.1 | NP_001264003.1 | ||
| NM_144716.5 | NP_653317.2 | |||
| NR_102269.1 | ||||
| XM_011533390.2 | XP_011531692.1 | |||
| XM_011533391.2 | XP_011531693.1 | |||
| XM_011533392.2 | XP_011531694.1 | |||
| XM_011533394.2 | XP_011531696.1 | |||
| XM_011533396.2 | XP_011531698.1 | |||
| AK096749.1 | BAC04857.1 | |||
| AK295532.1 | ||||
| AK303160.1 | ||||
| BC020830.1 | AAH20830.1 | |||
| BM803652.1 | ||||
| CN430023.1 | ||||
| CV574708.1 | ||||
| DC379144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3596071 | Chr.3:46919793 - 46948500 on Build GRCh38 | Loss |
|
| nsv3800 | Chr.3:46920158 - 46965751 on Build GRCh38 | Deletion |
|
| esv3596074 | Chr.3:46936636 - 46948496 on Build GRCh38 | Loss |
|
| nsv460526 | Chr.3:46894037 - 46959860 on Build GRCh38 | Loss |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
| esv3596073 | Chr.3:46936296 - 47131646 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map