Assay Details
Target Gene Details
Entrez Gene ID: | 151742 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1L |
Gene Aliases: |
PP2C-epsilon, PP2CE, PPM1-LIKE |
Location: |
Chr.3:160755532-161078907 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1L | NM_139245.3 | NP_640338.2 | ||
| XM_011512440.2 | XP_011510742.1 | |||
| AY337264.1 | AAR00269.1 | |||
| DB032505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598389 | Chr.3:160767086 - 160857900 on Build GRCh38 | Loss |
|
| esv3598390 | Chr.3:160768581 - 160829645 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map