Assay Details
Target Gene Details
Entrez Gene ID: | 84319 |
Gene Name: | cms1 ribosomal small subunit homolog (yeast) |
Gene Aliases: |
C3orf26 |
Location: |
Chr.3:99817834-100178632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMSS1 | NM_032359.3 | NP_115735.2 | ||
AK124387.1 | ||||
AK291392.1 | ||||
AK312483.1 | ||||
BC006475.1 | AAH06475.1 | |||
BC006512.2 | AAH06512.1 |
Target Gene Details
Entrez Gene ID: | 101929357 |
Gene Name: | uncharacterized LOC101929357 |
Gene Aliases: |
- |
Location: |
Chr.3:99816632-99823865 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HP09053 | NR_110821.1 | |||
AB593151.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
