Assay Details
Target Gene Details
Entrez Gene ID: | 5922 |
Gene Name: | RAS p21 protein activator 2 |
Gene Aliases: |
GAP1M |
Location: |
Chr.3:141487047-141615363 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RASA2 | NM_001303245.1 | NP_001290174.1 | ||
NM_001303246.1 | NP_001290175.1 | |||
NM_006506.3 | NP_006497.2 | |||
XM_011513059.2 | XP_011511361.1 | |||
XM_017006968.1 | XP_016862457.1 | |||
XM_017006969.1 | XP_016862458.1 | |||
XM_017006970.1 | XP_016862459.1 | |||
XM_017006971.1 | XP_016862460.1 | |||
XM_017006972.1 | XP_016862461.1 | |||
AF115573.1 | AAD09821.1 | |||
AK292016.1 | ||||
AK301015.1 | ||||
D78155.1 | BAA11230.1 | |||
D82880.1 | BAA11621.1 | |||
DA792257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1006237 | Chr.3:141508683 - 141581880 on Build GRCh38 | Loss |
![]() |
esv2676781 | Chr.3:141519540 - 141526900 on Build GRCh38 | Deletion |
![]() |
esv3597979 | Chr.3:141519480 - 141526831 on Build GRCh38 | Loss |
![]() |
nsv1014365 | Chr.3:141516537 - 141563773 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
