Assay Details
Target Gene Details
Entrez Gene ID: | 5352 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 2 |
Gene Aliases: |
BRKS2, LH2, TLH |
Location: |
Chr.3:146069436-146161495 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLOD2 | NM_000935.2 | NP_000926.2 | ||
NM_182943.2 | NP_891988.1 | |||
XM_005247535.3 | XP_005247592.1 | |||
XM_017006625.1 | XP_016862114.1 | |||
AB209879.1 | BAD93116.1 | |||
AK125700.1 | ||||
AK295084.1 | ||||
AL832238.1 | ||||
BC037169.1 | AAH37169.1 | |||
BX537521.1 | ||||
U84573.1 | AAB58363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525829 | Chr.3:145896530 - 146145992 on Build GRCh38 | Loss |
![]() |
nsv4046 | Chr.3:145975462 - 146162402 on Build GRCh38 | Deletion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
