Assay Details
Target Gene Details
Entrez Gene ID: | 6476 |
Gene Name: | sucrase-isomaltase |
Gene Aliases: |
- |
Location: |
Chr.3:164978898-165078495 on Build GRCh38 |
Assay Gene Location: | Within Intron 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SI | NM_001041.3 | NP_001032.2 | ||
| XM_011513078.2 | XP_011511380.1 | |||
| BC115034.1 | AAI15035.1 | |||
| BC116452.1 | ||||
| BC132834.1 | ||||
| BC132860.1 | ||||
| X63597.1 | CAA45140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575627 | Chr.3:164978711 - 165081291 on Build GRCh38 | Gain |
|
| nsv1014800 | Chr.3:164983237 - 165073353 on Build GRCh38 | Gain |
|
| nsv592446 | Chr.3:164547652 - 165237953 on Build GRCh38 | Loss |
|
| nsv428426 | Chr.3:164382966 - 165879735 on Build GRCh38 | Loss |
|
| nsv829785 | Chr.3:164860796 - 164996990 on Build GRCh38 | Loss |
|
| dgv8717n54 | Chr.3:164935636 - 165075970 on Build GRCh38 | Loss |
|
| esv2763324 | Chr.3:164978854 - 165073365 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112617507] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map