Assay Details
Target Gene Details
Entrez Gene ID: | 26137 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
DPZF, HOF, ODA-8S, PRIMS, ZNF288 |
Location: |
Chr.3:114314500-115147280 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB20 | NM_015642.5 | NP_056457.3 | ||
BX647778.1 | CAH56190.1 | |||
BX648237.1 |
Target Gene Details
Entrez Gene ID: | 100874131 |
Gene Name: | ZBTB20 antisense RNA 4 |
Gene Aliases: |
- |
Location: |
Chr.3:115100423-115103061 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB20-AS4 | NR_046877.1 | |||
DA196340.1 | ||||
DA301952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv955327 | Chr.3:115100154 - 115146253 on Build GRCh38 | Duplication |
![]() ![]() |
nsv829683 | Chr.3:114973157 - 115149982 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
