Assay Details
Target Gene Details
Entrez Gene ID: | 23395 |
Gene Name: | leucyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
LEURS, PRLTS4, mtLeuRS |
Location: |
Chr.3:45388561-45548842 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARS2 | NM_015340.3 | NP_056155.1 | ||
| XM_005265006.1 | XP_005265063.1 | |||
| XM_011533554.2 | XP_011531856.1 | |||
| XM_017006042.1 | XP_016861531.1 | |||
| AK025465.1 | ||||
| AK026113.1 | ||||
| AK308146.1 | ||||
| BC025989.2 | AAH25989.1 | |||
| D21851.1 |
Target Gene Details
Entrez Gene ID: | 100885795 |
Gene Name: | LARS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:45482695-45509545 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARS2-AS1 | NR_048543.1 | |||
| DB017595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8301n54 | Chr.3:45507536 - 45509980 on Build GRCh38 | Loss |
|
| esv25561 | Chr.3:45501795 - 45510572 on Build GRCh38 | Loss |
|
| dgv8303n54 | Chr.3:45507806 - 45514461 on Build GRCh38 | Loss |
|
| nsv590177 | Chr.3:45508299 - 45509915 on Build GRCh38 | Loss |
|
| esv3561814 | Chr.3:45501509 - 45510579 on Build GRCh38 | Deletion |
|
| esv2664866 | Chr.3:45501801 - 45510566 on Build GRCh38 | Deletion |
|
| nsv834677 | Chr.3:45343270 - 45511215 on Build GRCh38 | Gain |
|
| nsv590162 | Chr.3:45500591 - 45510525 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs374367931] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map