Assay Details
Target Gene Details
Entrez Gene ID: | 90102 |
Gene Name: | pleckstrin homology like domain family B member 2 |
Gene Aliases: |
LL5b, LL5beta |
Location: |
Chr.3:111732480-111976517 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHLDB2 | NM_001134437.1 | NP_001127909.1 | ||
AK296841.1 | ||||
AK307491.1 | ||||
CR749654.1 | CAH18447.1 | |||
DA447763.1 |
Target Gene Details
Entrez Gene ID: | 257068 |
Gene Name: | phosphatidylinositol specific phospholipase C X domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:111674676-111846447 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCXD2 | NM_001185106.1 | NP_001172035.1 | ||
NM_153268.3 | NP_695000.1 | |||
AK056141.1 | BAB71103.1 | |||
BC121155.1 | ||||
BC121156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460821 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv591290 | Chr.3:111708608 - 111743723 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
