Assay Details
Target Gene Details
Entrez Gene ID: | 2549 |
Gene Name: | GRB2 associated binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.4:143336830-143474565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAB1 | NM_002039.3 | NP_002030.2 | ||
| NM_207123.2 | NP_997006.1 | |||
| XM_017007969.1 | XP_016863458.1 | |||
| XM_017007970.1 | XP_016863459.1 | |||
| XM_017007971.1 | XP_016863460.1 | |||
| AK289767.1 | ||||
| AK314635.1 | ||||
| BC064848.1 | AAH64848.1 | |||
| DA109618.1 | ||||
| U43885.1 | AAC50380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5411n100 | Chr.4:143367285 - 143378410 on Build GRCh38 | Gain |
|
| nsv1018566 | Chr.4:143363774 - 143377777 on Build GRCh38 | Loss |
|
| esv3602506 | Chr.4:143377287 - 143378637 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map