Assay Details
Target Gene Details
Entrez Gene ID: | 23022 |
Gene Name: | palladin, cytoskeletal associated protein |
Gene Aliases: |
CGI-151, CGI151, MYN, PNCA1, SIH002 |
Location: |
Chr.4:168497039-168928457 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALLD | NM_001166108.1 | NP_001159580.1 | ||
NM_016081.3 | NP_057165.3 | |||
XM_005262861.4 | XP_005262918.1 | |||
XM_011531768.2 | XP_011530070.1 | |||
XM_011531769.2 | XP_011530071.1 | |||
XM_011531770.2 | XP_011530072.1 | |||
XM_011531771.2 | XP_011530073.1 | |||
XM_011531772.2 | XP_011530074.1 | |||
XM_011531773.1 | XP_011530075.1 | |||
XM_011531774.1 | XP_011530076.1 | |||
XM_017007910.1 | XP_016863399.1 | |||
XM_017007911.1 | XP_016863400.1 | |||
AF464873.1 | AAL69964.1 | |||
AK096458.1 | BAC04796.1 | |||
BC140850.1 | ||||
BC144666.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
