Assay Details
Target Gene Details
Entrez Gene ID: | 11275 |
Gene Name: | kelch like family member 2 |
Gene Aliases: |
ABP-KELCH, MAV, MAYVEN |
Location: |
Chr.4:165207618-165323156 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL2 | NM_001161521.1 | NP_001154993.1 | ||
| NM_001161522.1 | NP_001154994.1 | |||
| NM_007246.3 | NP_009177.3 | |||
| XM_011531572.2 | XP_011529874.1 | |||
| XM_011531575.2 | XP_011529877.1 | |||
| XM_011531576.2 | XP_011529878.1 | |||
| XM_017007674.1 | XP_016863163.1 | |||
| XM_017007675.1 | XP_016863164.1 | |||
| XM_017007676.1 | XP_016863165.1 | |||
| XM_017007677.1 | XP_016863166.1 | |||
| AB209727.1 | BAD92964.1 | |||
| AF059569.1 | AAC67502.1 | |||
| AK025289.1 | ||||
| AK294103.1 | ||||
| AK294336.1 | ||||
| AK308940.1 | ||||
| AK315372.1 | ||||
| BC022503.1 | AAH22503.1 | |||
| BC036468.2 | AAH36468.1 | |||
| BX647776.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73860632] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map