Assay Details
Target Gene Details
Entrez Gene ID: | 7368 |
Gene Name: | UDP glycosyltransferase 8 |
Gene Aliases: |
CGT, UGT4 |
Location: |
Chr.4:114597789-114687914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT8 | NM_001128174.2 | NP_001121646.1 | ||
| NM_001322112.1 | NP_001309041.1 | |||
| NM_001322113.1 | NP_001309042.1 | |||
| NM_001322114.1 | NP_001309043.1 | |||
| AK127970.1 | ||||
| AK225763.1 | ||||
| BC075069.2 | AAH75069.1 | |||
| BP219039.1 | ||||
| DA295156.1 | ||||
| DC335502.1 | ||||
| U62899.1 | AAC51187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4478 | Chr.4:114575841 - 114607995 on Build GRCh38 | Deletion |
|
| esv2759275 | Chr.4:114420923 - 114669169 on Build GRCh38 | Loss |
|
| esv2752021 | Chr.4:114368240 - 114698514 on Build GRCh38 | Gain |
|
| esv3601829 | Chr.4:114512988 - 114663121 on Build GRCh38 | Gain |
|
| nsv1002816 | Chr.4:114411140 - 114611220 on Build GRCh38 | Gain |
|
| esv3601835 | Chr.4:114600577 - 114660419 on Build GRCh38 | Gain |
|
| nsv524515 | Chr.4:113918114 - 114701151 on Build GRCh38 | Gain |
|
| nsv830039 | Chr.4:114559965 - 114747581 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map