Assay Details
Target Gene Details
Entrez Gene ID: | 7469 |
Gene Name: | negative elongation factor complex member A |
Gene Aliases: |
NELF-A, P/OKcl.15, WHSC2 |
Location: |
Chr.4:1982714-2009972 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELFA | NM_005663.4 | NP_005654.3 | ||
| XM_017008589.1 | XP_016864078.1 | |||
| AB044549.1 | BAB18651.1 | |||
| AF101434.1 | AAC72982.1 | |||
| AF131751.1 | AAD20034.1 | |||
| AK001304.1 | ||||
| AK094040.1 | ||||
| AK095737.1 | ||||
| AK126056.1 | ||||
| AK222853.1 | BAD96573.1 | |||
| BC002764.2 | AAH02764.2 |
More Information
Set Membership: |
|

Genomic Map