Assay Details
Target Gene Details
Entrez Gene ID: | 55351 |
Gene Name: | serine/threonine kinase 32B |
Gene Aliases: |
HSA250839, STK32, STKG6, YANK2 |
Location: |
Chr.4:5051323-5501001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK32B | NM_001306082.1 | NP_001293011.1 | ||
| NM_018401.2 | NP_060871.1 | |||
| XM_011513497.1 | XP_011511799.1 | |||
| AJ250839.1 | CAB76471.1 | |||
| AK292696.1 | ||||
| AK313846.1 | ||||
| BC038238.1 | AAH38238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5094n100 | Chr.4:4920733 - 5083483 on Build GRCh38 | Gain |
|
| nsv593479 | Chr.4:4899358 - 5459331 on Build GRCh38 | Gain |
|
| nsv997889 | Chr.4:4785891 - 5220458 on Build GRCh38 | Gain |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77513956] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map