Assay Details
Target Gene Details
Entrez Gene ID: | 1182 |
Gene Name: | chloride voltage-gated channel 3 |
Gene Aliases: |
CLC3, ClC-3 |
Location: |
Chr.4:169620521-169723187 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN3 | NM_001243372.1 | NP_001230301.1 | ||
| NM_001829.3 | NP_001820.2 | |||
| NM_173872.3 | NP_776297.2 | |||
| AB019542.1 | BAC54560.1 | |||
| AF029346.1 | AAB95161.1 | |||
| AF172729.1 | AAD51034.1 | |||
| AK304362.1 | ||||
| BC054877.1 | AAH54877.1 | |||
| BC136510.1 | ||||
| BX647119.1 | ||||
| X78520.1 | CAA55280.1 |
More Information
Set Membership: |
|

Genomic Map