Assay Details
Target Gene Details
Entrez Gene ID: | 79658 |
Gene Name: | Rho GTPase activating protein 10 |
Gene Aliases: |
GRAF2, PS-GAP, PSGAP |
Location: |
Chr.4:147732063-148072781 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP10 | NM_024605.3 | NP_078881.3 | ||
| XM_005263215.3 | XP_005263272.1 | |||
| XM_017008602.1 | XP_016864091.1 | |||
| AB050785.1 | BAB61771.1 | |||
| AK123785.1 | ||||
| BC109029.2 | AAI09030.1 | |||
| BC109030.1 | AAI09031.1 | |||
| BC126899.1 | ||||
| BC128055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602608 | Chr.4:147742992 - 147755464 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map