Assay Details
Target Gene Details
Entrez Gene ID: | 353497 |
Gene Name: | DNA polymerase nu |
Gene Aliases: |
POL4P |
Location: |
Chr.4:2071958-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLN | NM_181808.3 | NP_861524.2 | ||
| AF044578.1 | AAD02338.1 | |||
| AL122048.1 | ||||
| AY136549.1 | AAN52116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv949757 | Chr.4:2056974 - 2082173 on Build GRCh38 | Deletion |
|
| nsv822441 | Chr.4:1991240 - 2080440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map