Assay Details
Target Gene Details
Entrez Gene ID: | 101927186 |
Gene Name: | adhesion G protein-coupled receptor L3 antisense RNA 1 |
Gene Aliases: |
LPHN3-AS1 |
Location: |
Chr.4:62071755-62161765 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL3-AS1 | NR_110595.1 | |||
| DB518756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594332 | Chr.4:62111038 - 62168356 on Build GRCh38 | Loss |
|
| nsv1005620 | Chr.4:62112397 - 62169669 on Build GRCh38 | Loss |
|
| esv2611251 | Chr.4:62117199 - 62118132 on Build GRCh38 | Insertion |
|

Genomic Map