Assay Details
Target Gene Details
Entrez Gene ID: | 64854 |
Gene Name: | ubiquitin specific peptidase 46 |
Gene Aliases: |
- |
Location: |
Chr.4:52590960-52659335 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP46 | NM_001134223.1 | NP_001127695.1 | ||
NM_001286767.1 | NP_001273696.1 | |||
NM_001286768.1 | NP_001273697.1 | |||
NM_022832.3 | NP_073743.2 | |||
XM_017008557.1 | XP_016864046.1 | |||
AK022614.1 | BAB14133.1 | |||
AK024318.1 | BAB14881.1 | |||
AK296493.1 | ||||
AK299883.1 | ||||
AK302438.1 | ||||
BC037574.1 | AAH37574.1 | |||
GU455414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524494 | Chr.4:52593454 - 52715898 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv594201 | Chr.4:51932458 - 52866278 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
