Assay Details
Target Gene Details
Entrez Gene ID: | 166647 |
Gene Name: | adhesion G protein-coupled receptor A3 |
Gene Aliases: |
GPR125, PGR21, TEM5L |
Location: |
Chr.4:22387374-22516058 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA3 | NM_145290.3 | NP_660333.2 | ||
| XM_011513811.2 | XP_011512113.1 | |||
| XM_017007834.1 | XP_016863323.1 | |||
| AK027494.1 | ||||
| AK095866.1 | ||||
| AK299168.1 | ||||
| AL832257.1 | ||||
| AY181243.1 | AAO27355.1 | |||
| BC035645.1 | AAH35645.1 | |||
| BX648287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599899 | Chr.4:22389216 - 22392058 on Build GRCh38 | Loss |
|
| esv2656963 | Chr.4:22389125 - 22392022 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map