Assay Details
Target Gene Details
Entrez Gene ID: | 3360 |
Gene Name: | 5-hydroxytryptamine receptor 4 |
Gene Aliases: |
5-HT4, 5-HT4R |
Location: |
Chr.5:148451032-148654527 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR4 | NM_001040169.2 | NP_001035259.1 | ||
| NM_199453.3 | NP_955525.1 | |||
| AJ011371.1 | CAA09600.1 | |||
| AJ131725.1 | CAC79533.1 | |||
| AJ278979.1 | CAC22248.1 | |||
| AJ278981.1 | CAC22250.1 | |||
| AJ633645.1 | CAG17627.1 | |||
| Y08756.1 | CAA70002.1 | |||
| Y09586.1 | CAA70774.1 | |||
| Y13584.1 | CAA73911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607115 | Chr.5:148457429 - 148470982 on Build GRCh38 | Loss |
|
| nsv830516 | Chr.5:148421541 - 148586530 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map