Assay Details
Target Gene Details
Entrez Gene ID: | 153769 |
Gene Name: | SH3 domain containing ring finger 2 |
Gene Aliases: |
HEPP1, POSHER, PPP1R39, RNF158 |
Location: |
Chr.5:145936563-146081773 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF2 | NM_152550.3 | NP_689763.3 | ||
| XM_005268380.2 | XP_005268437.1 | |||
| XM_006714757.1 | XP_006714820.1 | |||
| XM_011537567.2 | XP_011535869.1 | |||
| XM_011537568.2 | XP_011535870.1 | |||
| XM_011537569.2 | XP_011535871.1 | |||
| XM_017009136.1 | XP_016864625.1 | |||
| AK074234.1 | BAB85025.1 | |||
| AK292576.1 | ||||
| AL833297.1 | ||||
| BC125106.1 | ||||
| BC125107.1 |
More Information
Set Membership: |
|

Genomic Map