Assay Details
Target Gene Details
Entrez Gene ID: | 64374 |
Gene Name: | SIL1 nucleotide exchange factor |
Gene Aliases: |
BAP, MSS, ULG5 |
Location: |
Chr.5:138946720-139198376 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIL1 | NM_001037633.1 | NP_001032722.1 | ||
| NM_022464.4 | NP_071909.1 | |||
| XM_011543570.2 | XP_011541872.1 | |||
| AF547994.1 | AAN84477.1 | |||
| AJ299442.1 | CAC17773.1 | |||
| AK074624.1 | BAC11096.1 | |||
| AK075177.1 | BAC11452.1 | |||
| AY358950.1 | AAQ89309.1 | |||
| BC011568.1 | AAH11568.1 | |||
| KU178743.1 | ||||
| KU178744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950408 | Chr.5:138944112 - 138956011 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map