Assay Details
Target Gene Details
Entrez Gene ID: | 101926904 |
Gene Name: | long intergenic non-protein coding RNA 1265 |
Gene Aliases: |
- |
Location: |
Chr.5:38710367-38720273 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01265 | NR_104631.1 | |||
| AA393775.1 |
Target Gene Details
Entrez Gene ID: | 101929768 |
Gene Name: | OSMR antisense RNA 1 (head to head) |
Gene Aliases: |
CTD-2127H9.1 |
Location: |
Chr.5:38693213-38845829 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSMR-AS1 | NR_109951.1 | |||
| DA342801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604771 | Chr.5:38622860 - 38816266 on Build GRCh38 | Gain |
|
| esv3604761 | Chr.5:38056637 - 38805862 on Build GRCh38 | Gain |
|

Genomic Map