Assay Details
Target Gene Details
Entrez Gene ID: | 101927075 |
Gene Name: | FGF10 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:44388732-44413989 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF10-AS1 | NR_108034.1 | 3 | 1273 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830277 | Chr.5:44383300 - 44566779 on Build GRCh38 | Gain |
|

Genomic Map