Assay Details
Target Gene Details
Entrez Gene ID: | 1022 |
Gene Name: | cyclin dependent kinase 7 |
Gene Aliases: |
CAK, CAK1, CDKN7, HCAK, MO15, STK1, p39MO15 |
Location: |
Chr.5:69234795-69277430 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK7 | NM_001324069.1 | NP_001310998.1 | ||
| NM_001324070.1 | NP_001310999.1 | |||
| NM_001324071.1 | NP_001311000.1 | |||
| NM_001324072.1 | NP_001311001.1 | |||
| NM_001324074.1 | NP_001311003.1 | |||
| NM_001324075.1 | NP_001311004.1 | |||
| NM_001324077.1 | NP_001311006.1 | |||
| NM_001324078.1 | NP_001311007.1 | |||
| NM_001799.3 | NP_001790.1 | |||
| NR_136690.1 | ||||
| XM_011543094.2 | XP_011541396.1 | |||
| AK026509.1 | ||||
| AK308493.1 | ||||
| BC000834.2 | AAH00834.1 | |||
| BC005298.1 | AAH05298.1 | |||
| BI520184.1 | ||||
| BU660651.1 | ||||
| KU177970.1 | ||||
| KU177971.1 | ||||
| L20320.1 | AAA36657.1 | |||
| X77303.1 | CAA54508.1 | |||
| X77743.1 | CAA54793.1 | |||
| X79193.1 | CAA55785.1 | |||
| Y13120.1 | CAA73587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map