Assay Details
Target Gene Details
Entrez Gene ID: | 51808 |
Gene Name: | phosphorylated adaptor for RNA export |
Gene Aliases: |
RNUXA |
Location: |
Chr.5:126600915-126627252 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHAX | NM_032177.3 | NP_115553.2 | ||
| AK023255.1 | BAB14489.1 | |||
| AK024065.1 | BAB14809.1 | |||
| AK225464.1 | ||||
| AK298497.1 | ||||
| BC021161.2 | AAH21161.1 | |||
| CR457305.1 | CAG33586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462442 | Chr.5:126586076 - 126753498 on Build GRCh38 | Gain |
|
| nsv1030016 | Chr.5:126553411 - 126636590 on Build GRCh38 | Gain |
|
| esv3606679 | Chr.5:126424914 - 126763280 on Build GRCh38 | Gain |
|
| esv3606680 | Chr.5:126475605 - 126628219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map